2,6-Dimethylhydroquinone
Supplier: Matrix Scientific
Synonyms:
2,5-Dihydroxy-m-xylene
004077-1G
004077-5G
004077-25G
101777-322EA
104.62
USD
101777-322
101777-320
101777-324
2,6-Dimethylhydroquinone
2,6-Dimethylhydroquinone
TCA listed: T
Hazard: Toxic/Irritant
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Melting Pt: 149…150 °C |
MDL Number:
MFCD00016466 CAS Number: 654-42-2 |
Specification Test Results
Melting point | 149-150° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...