2,3-Dimethylbenzylamine ≥98%
Supplier: Matrix Scientific
004073-1G
004073-5G
101777-314EA
81.54
USD
101777-314
101777-312
2,3-Dimethylbenzylamine ≥98%
2,3-Dimethylbenzylamine
Hazard: Corrosive/Air Sensitive
Formula:
C₉H₁₃N MW: 135.21 g/mol Boiling Pt: 101…103 °C (15 mmHg) Density: 0.97 |
MDL Number:
MFCD00052680 CAS Number: 51586-20-0 |
Specification Test Results
Boiling point | 101-103°/1 |
Density | 0.97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...