2-Bromoethyl isothiocyanate ≥98%

Supplier: Matrix Scientific

004040-5G
101777-306EA 253.85 USD
101777-306
2-Bromoethyl isothiocyanate ≥98%
2-Bromoethyl isothiocyanate

Hazard: Toxic/Corrosive/Store in Freezer

Formula: BrH₂CCH₂NCS
MW: 166.04 g/mol
Boiling Pt: 102…108 °C (15 mmHg)
Density: 1.65
MDL Number: MFCD00060415
CAS Number: 1483-41-6

Order Now

Specification Test Results

Boiling point 102-108°/1
Density 1.65

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR