2-Chlorothiophenol ≥99%
Supplier: Matrix Scientific
Synonyms:
o-Chlorothiophenol
, 1-Chloro-2-mercaptobenzene
, 2-Chlorobenzene-1-thiol
, 2-Chlorobenzenethiol
, 2-chlorothiophenol
003972-25G
003972-100G
101777-278EA
269.23
USD
101777-278
101777-276
2-Chlorothiophenol ≥99%
2-Chlorothiophenol
TCA listed: T
Hazard: Corrosive/Stench
Formula:
C₆H₅ClS MW: 144.62 g/mol Boiling Pt: 205…206 °C Density: 1.275 |
MDL Number:
MFCD00004830 CAS Number: 6320-03-2 |
Specification Test Results
Boiling point | 205-206°/2 |
Density | 1.275 |
ND20 | 1.6029 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...