3-Chlorothiophene ≥97%
Supplier: Matrix Scientific
003961-25G
003961-100G
101777-266EA
181.54
USD
101777-266
101777-264
3-Chlorothiophene ≥97%
3-Chlorothiophene
Hazard: Flammable/Stench
Formula:
C₄H₃ClS MW: 118.59 g/mol Boiling Pt: 137…139 °C Density: 1.276 |
MDL Number:
MFCD00043887 CAS Number: 17249-80-8 UN: 1992 ADR: 3,III |
Specification Test Results
Boiling point | 137-139° |
Density | 1.276 |
ND20 | 1.553 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...