2-Chlorothiophene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Thienyl chloride

003960-500G
101777-262EA 218.46 USD
101777-262
2-Chlorothiophene ≥98%
2-Chlorothiophene

TCA listed: T
Hazard: Flammable/Toxic/Stench

Formula: C₄H₃ClS
MW: 118.59 g/mol
Boiling Pt: 127…129 °C
Density: 1.286
Flash Pt: 22 °C
MDL Number: MFCD00005421
CAS Number: 96-43-5
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Boiling point 127-129°
Density 1.286
ND20 1.548

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR