4-Benzylphenylacetonitrile ≥98%
Supplier: Matrix Scientific
003894-1G
003894-5G
101777-216EA
246.15
USD
101777-216
101777-214
4-Benzylphenylacetonitrile ≥98%
4-Benzylphenylacetonitrile
Hazard: Irritant
Formula:
C₁₅H₁₃N Melting Pt: 34…36 °C |
UN:
3439 ADR: 6.1,III |
Specification Test Results
Melting point | 34-36° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...