3,3,3-Trifluoropropene ≥99%
Supplier: Matrix Scientific
003813-100G
101777-140EA
515.38
USD
101777-140
3,3,3-Trifluoropropene ≥99%
3,3,3-Trifluoropropene
TCA listed: T
Hazard: Flammable Gas
Formula:
C₃H₃F₃ MW: 96.05 g/mol Boiling Pt: –18…–16 °C |
MDL Number:
MFCD00039265 CAS Number: 677-21-4 UN: 1954 ADR: 2, |
Specification Test Results
Boiling point | -18 to -16 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...