1,1,1-Trifluoropropane ≥97%

Supplier: Matrix Scientific

003812-25G
101777-138EA 1053.85 USD
101777-138
1,1,1-Trifluoropropane ≥97%
1,1,1-Trifluoropropane

Hazard: Flammable Gas

Formula: C₃H₅F₃
Boiling Pt: –12 °C
Melting Pt: –148 °C
CAS Number: 421-07-8

Order Now

Specification Test Results

Boiling point -12°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR