Ethyl nonafluorovalerate ≥98%

Supplier: Matrix Scientific

Synonyms: Nonafluorovaleric Acid Ethyl EsterPerfluorovaleric Acid Ethyl EsterEthyl Perfluorovalerate

003772-100G 003772-25G
101777-112EA 530.77 USD
101777-112 101777-110
Ethyl nonafluorovalerate ≥98%
Ethyl nonafluorovalerate

Purity: 98%
CAS: 424-36-2
MDL Number: MFCD00077523
Molecular Formula: C7H5F9O2
Molecular Weight: 292,1
Boiling Point: 121°C
Hazard: Flammable

Formula: C₇H₅F₉O₂
MW: 292.10 g/mol
Boiling Pt: 121 °C
MDL Number: MFCD00077523
CAS Number: 424-36-2

Order Now

Specification Test Results

Boiling point 121°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR