1,2,3-Trifluoro-4-nitrobenzene ≥99%

Supplier: Matrix Scientific

Synonyms: 2,3,4-Trifluoronitrobenzene

003762-100G
101777-100EA 44.62 USD
101777-100
1,2,3-Trifluoro-4-nitrobenzene ≥99%
1,2,3-Trifluoro-4-nitrobenzene

Hazard: Toxic

Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 92 °C (20 mmHg)
Density: 1.541
MDL Number: MFCD00041546
CAS Number: 771-69-7
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 92°/20mm
Density 1.541
ND20 1.492

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR