N-[3-(Trifluoromethyl)phenyl]benzamide ≥98%

Supplier: Matrix Scientific

003745-1G
101777-086EA 267.69 USD
101777-086
N-[3-(Trifluoromethyl)phenyl]benzamide ≥98%
N-[3-(Trifluoromethyl)phenyl]benzamide

Hazard: Irritant

Formula: C₁₄H₁₀F₃NO
Melting Pt: 107…109 °C
MDL Number: MFCD00049013
CAS Number: 1939-24-8

Order Now

Specification Test Results

Melting point 107-109°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR