4,4,4-Trifluorocrotonamide ≥95%
Supplier: Matrix Scientific
003693-5G
003693-25G
003693-1G
101777-034EA
453.85
USD
101777-034
101777-032
101777-030
4,4,4-Trifluorocrotonamide ≥95%
4,4,4-Trifluorocrotonamide
Hazard: Irritant/Moisture Sensitive
Formula:
C₄H₄F₃NO Melting Pt: 147…150 °C |
MDL Number:
MFCD00190644 CAS Number: 590-76-1 |
Specification Test Results
Melting point | 147-150° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...