1,2,4-Trifluorobenzene ≥98%

Supplier: Matrix Scientific

003679-100G
101777-002EA 95.38 USD
101777-002
1,2,4-Trifluorobenzene ≥98%
1,2,4-Trifluorobenzene

Hazard: Flammable

Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 88 °C (759 mmHg)
Density: 1.264
MDL Number: MFCD00000305
CAS Number: 367-23-7
UN: 2387
ADR: 3,II

Order Now

Specification Test Results

Boiling point 88°/759mm
Density 1.264
ND20 1.423

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR