2,3,6-Trifluorobenzoyl chloride ≥98%
Supplier: Matrix Scientific
003678-1G
003678-5G
101777-000EA
96.92
USD
101777-000
101776-998
2,3,6-Trifluorobenzoyl chloride ≥98%
2,3,6-Trifluorobenzoyl chloride
Hazard: Corrosive
Formula:
C₇H₂ClF₃O MW: 194.54 g/mol Boiling Pt: 77…79 °C (20 mmHg) Density: 1.48 |
MDL Number:
MFCD00061203 CAS Number: 189807-20-3 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 77-79°/20m |
Density | 1.48 |
ND20 | 1.483 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...