2,3,4-Trifluorobenzoyl chloride ≥98%
Supplier: Matrix Scientific
Synonyms:
2,3,4-trifluorobenzoylchloride
003671-5G
003671-1G
101776-980EA
40
USD
101776-980
101776-982
2,3,4-Trifluorobenzoyl chloride ≥98%
2,3,4-Trifluorobenzoyl chloride
Hazard: Corrosive
Formula:
C₇H₂ClF₃O MW: 194.54 g/mol Boiling Pt: 143…144 °C (130 mmHg) Density: 1.522 Flash Pt: 92 °C |
MDL Number:
MFCD00075078 CAS Number: 157373-08-5 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 143-144°/1 |
Density | 1.522 |
ND20 | 1.497 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...