1,1,1,4,4,4-Hexafluoro-2-butene mixture E/Z
Supplier: Matrix Scientific
001235-25G
001235-100G
001235-5G
101775-268EA
153.85
USD
101775-268
101775-272
101775-270
1,1,1,4,4,4-Hexafluoro-2-butene mixture E/Z
1,1,1,4,4,4-Hexafluoro-2-butene
Hazard: Nonflammable Gas/Refrigerate
Formula:
C₄H₂F₆ Boiling Pt: 8.5 °C |
MDL Number:
MFCD00798134 CAS Number: 407-60-3 |
Specification Test Results
Boiling point | 8.5° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...