2,2,3,3,4,4,4-Heptafluorobutyltrifluoromethanesulfonate ≥97%

Supplier: Matrix Scientific

001207-5G 001207-1G
101775-240EA 423.08 USD
101775-240 101775-274
2,2,3,3,4,4,4-Heptafluorobutyltrifluoromethanesulfonate ≥97%
2,2,3,3,4,4,4-Heptafluorobutyltrifluoromethanesulfonate

Hazard: Toxic/Corrosive

Formula: C₅H₂F₁₀O₃S
MW: 332.12 g/mol
Boiling Pt: 118…120 °C (730 mmHg)
MDL Number: MFCD02093339
CAS Number: 6401-01-0

Order Now

Specification Test Results

Boiling point 118-120°/7
ND20 1.304

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR