1,1,1,3,3,3-Hexafluoroisopropyl acrylate ≥99%
Supplier: Matrix Scientific
001197-25G
101775-236EA
89.23
USD
101775-236
1,1,1,3,3,3-Hexafluoroisopropyl acrylate ≥99%
1,1,1,3,3,3-Hexafluoroisopropyl acrylate
TCA listed: T
Hazard: Flammable/Irritant/Refrigerate
Formula:
C₆H₄F₆O₂ MW: 222.09 Boiling Pt: 84 °C Density: 1.33 |
UN:
3272 ADR: 3,II |
Specification Test Results
Boiling point | 84° |
Density | 1.33 |
ND20 | 1.319 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...