2-Bromo-2,3,3,3-tetrafluoropropionyl chloride ≥97%
Supplier: Matrix Scientific
001160-5G
001160-1G
101775-224EA
85.1
USD
101775-224
101775-222
2-Bromo-2,3,3,3-tetrafluoropropionyl chloride ≥97%
2-Bromo-2,3,3,3-tetrafluoropropionyl chloride
Hazard: Corrosive
Formula:
C₃BrClF₄O MW: 243.39 g/mol Boiling Pt: 46 °C |
MDL Number:
MFCD00153723 CAS Number: 6066-45-1 |
Specification Test Results
Boiling point | 67-70° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...