2-Bromo-1,1-difluoroethylene ≥98% inhibited

Supplier: Matrix Scientific


Stabilization: Stabilised with d-limonene

001134-25G
101775-210EA 669.23 USD
101775-210
2-Bromo-1,1-difluoroethylene ≥98% inhibited
2-Bromo-1,1-difluoroethylene

Inhibited with d-limonene


TCA listed: T
Hazard: Flammable Gas

Formula: C₂HBrF₂
Boiling Pt: 6 °C
Melting Pt: –115 °C
Density: 1.817/0.5 °C
CAS Number: 359-08-0
UN: 3161
ADR: 2,

Order Now

Specification Test Results

Boiling point

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR