2,4,6-Trifluorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Trifluorobenzene-1-sulfonyl chloride2,4,6-Trifluorobenzene-1-sulphonyl chloride

003667-5G 003667-1G
101776-970EA 112.31 USD
101776-970 101776-968
2,4,6-Trifluorobenzenesulfonyl chloride ≥97%
2,4,6-Trifluorobenzenesulphonyl chloride

Hazard: Corrosive

Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
MDL Number: MFCD01091015
CAS Number: 220239-64-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR