3,4,5-Trifluorobenzoyl chloride ≥97%

Supplier: Matrix Scientific

003641-1G 003641-5G
101776-920EA 44.62 USD
101776-920 101776-922
3,4,5-Trifluorobenzoyl chloride ≥97%
3,4,5-Trifluorobenzoyl chloride

Hazard: Corrosive

Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Melting Pt: 122…123 °C
MDL Number: MFCD00083526
CAS Number: 177787-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR