2,4,6-Trifluorobenzoyl chloride ≥98%

Supplier: Matrix Scientific

Synonyms: 2,4,6-trifluorobenzoylchloride

003640-5G 003640-25G
101776-916EA 152.31 USD
101776-916 101776-918
2,4,6-Trifluorobenzoyl chloride ≥98%
2,4,6-Trifluorobenzoyl chloride

Hazard: Corrosive

Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Boiling Pt: 167…168 °C
Density: 1.499
MDL Number: MFCD00070810
CAS Number: 79538-29-7
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR