1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione ≥95%

Supplier: Matrix Scientific

Synonyms: 5,5-Dimethyl-1,1,1-trifluorohexane-2,4-dione

003589-5G 003589-25G
101776-854EA 207.69 USD
101776-854 101776-856
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione ≥95%
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione

TCA listed: T
Hazard: Flammable

Formula: C₈H₁₁F₃O₂
MW: 196.17 g/mol
Boiling Pt: 68…71 °C (6 mmHg)
Density: 1.129
MDL Number: MFCD00040841
CAS Number: 22767-90-4
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Boiling point 68-71°/6mm
Density 1.129
ND20 1.4026

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR