1,1,1-Trifluoro-2,3-epoxybutane, Technical Grade
Supplier: Matrix Scientific
003564-5G
003564-1G
101776-822EA
143.08
USD
101776-822
101776-824
1,1,1-Trifluoro-2,3-epoxybutane, Technical Grade
1,1,1-Trifluoro-2,3-epoxybutane
Hazard: Flammable/Toxic
Formula:
C₄H₅F₃O Boiling Pt: 55…58 °C |
MDL Number:
MFCD00236721 CAS Number: 406-30-4 |
Specification Test Results
Boiling point | 55-58° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...