1,1,3,3-Tetrafluorodimethyl ether ≥97%

Supplier: Matrix Scientific

003492-25G
101776-762EA 796.92 USD
101776-762
1,1,3,3-Tetrafluorodimethyl ether ≥97%
1,1,3,3-Tetrafluorodimethyl ether

Hazard: Nonflammable Gas

Formula: C₂H₂F₄O
Boiling Pt: 4.7 °C
MDL Number: MFCD00042097
CAS Number: 1691-17-4

Order Now

Specification Test Results

Boiling point 4.7°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR