2,2,3,3-Tetrafluoropropyl trifluoromethanesulfonate ≥97%

003478-1G
101776-744EA 224.18 USD
101776-744
2,2,3,3-Tetrafluoropropyl trifluoromethanesulfonate ≥97%
2,2,3,3-Tetrafluoropropyl trifluoromethanesulfonate

Hazard: Toxic/Corrosive

Formula: C₄H₃F₇O₃S
MW: 264.12 g/mol
Boiling Pt: 122…125 °C (737 mmHg)
Melting Pt: 114…118 °C
MDL Number: MFCD02093338
CAS Number: 6401-02-1

Order Now

Specification Test Results

Boiling point 122-125°/7
ND20 1.3223

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR