1,2,2,2-Tetrafluoroethyl trifluoromethyl ether ≥97%

Supplier: Matrix Scientific

003474-5G
101776-738EA 336.92 USD
101776-738
1,2,2,2-Tetrafluoroethyl trifluoromethyl ether ≥97%
1,2,2,2-Tetrafluoroethyl trifluoromethyl ether

Hazard: Nonflammable Gas

Formula: C₃HF₇O
Boiling Pt: –9.4 °C
MDL Number: MFCD01320709
CAS Number: 2356-62-9

Order Now

Specification Test Results

Boiling point -9.4°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR