Dibromoacetonitrile ≥95%

Supplier: Matrix Scientific

003425-100G 003425-25G
101776-718EA 354.12 USD
101776-718 101776-716
Dibromoacetonitrile ≥95%
Dibromoacetonitrile

TCA listed: T
Hazard: Toxic/Lachrymator

Formula: Br₂CHCN
MW: 198.84 g/mol
Boiling Pt: 67…69 °C (24 mmHg)
Density: 2.296
MDL Number: MFCD00001856
CAS Number: 3252-43-5
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 67-69°/24m
Density 2.296
ND20 1.539

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR