Ethyl-2-methyl-4,4,4-trifluorocrotonate ≥95%

Supplier: Matrix Scientific

003395-5G 003395-1G
101776-690EA 56.92 USD
101776-690 101776-692
Ethyl-2-methyl-4,4,4-trifluorocrotonate ≥95%
Ethyl-2-methyl-4,4,4-trifluorocrotonate

Hazard: Flammable

Formula: C₇H₉F₃O₂
Boiling Pt: 134…135 °C
MDL Number: MFCD00190637
CAS Number: 128227-97-4

Order Now

Specification Test Results

Boiling point 134-135°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR