3-Fluorobenzyl isothiocyanate ≥97%

Supplier: Matrix Scientific

003338-1G 003338-5G
101776-654EA 35.38 USD
101776-654 101776-656
3-Fluorobenzyl isothiocyanate ≥97%
3-Fluorobenzyl isothiocyanate

Hazard: Toxic/Refrigerate/Lachrymator

Formula: C₈H₆FNS
MW: 167.21 g/mol
Boiling Pt: 138 °C (16 mmHg)
Density: 1.22
MDL Number: MFCD00060406
CAS Number: 63351-94-0

Order Now

Specification Test Results

Boiling point 138°/16mm
Density 1.22

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR