α-Bromo-3-fluorotoulene ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Fluorobenzyl bromide
, α-Bromo-3-fluorotoluene
, 1-(Bromomethyl)-3-fluorobenzene
, m-Fluorobenzyl bromide
, alpha-Bromo-m-fluorotoulene
, α-Bromo-m-fluorotoulene
003330-25G
003330-100G
101776-646EA
73.85
USD
101776-646
101776-648
α-Bromo-3-fluorotoulene ≥97%
α-Bromo-3-fluorotoulene
TCA listed: T
Hazard: Corrosive/Lachrymator
Formula:
C₇H₆BrF MW: 189.03 g/mol Boiling Pt: 88 °C (20 mmHg) Density: 1.541 |
MDL Number:
MFCD00000340 CAS Number: 456-41-7 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 88°/20mm |
Density | 1.541 |
ND20 | 1.546 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...