α,α,α,3-Tetrafluorotoluene ≥99%
Supplier: Matrix Scientific
Synonyms:
3-Fluorobenzotrifluoride
, 1-Fluoro-3-(trifluoromethyl)benzene
003306-100G
003306-25G
101776-596EA
30.77
USD
101776-596
101776-594
α,α,α,3-Tetrafluorotoluene ≥99%
α,α,α,3-Tetrafluorotoluene
Hazard: Flammable
Formula:
C₇H₄F₄ MW: 164.1 g/mol Boiling Pt: 101…102 °C Density: 1.302 |
MDL Number:
MFCD00000382 CAS Number: 401-80-9 UN: 1993 ADR: 3,II |
Specification Test Results
Boiling point | 101-102° |
Density | 1.302 |
ND20 | 1.4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...