3-Cyanophenyl isothiocyanate ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Cyano-3-isothiocyanatobenzene
, 3-Isothiocyanatobenzonitrile
003209-5G
101776-538EA
232.31
USD
101776-538
3-Cyanophenyl isothiocyanate ≥98%
3-Cyanophenyl isothiocyanate
Hazard: Toxic/Corrosive/Moisture Sensitive/Refrigerate
Formula:
C₈H₄N₂S MW: 160.2 g/mol Melting Pt: 64…66 °C |
MDL Number:
MFCD00041070 CAS Number: 3125-78-8 UN: 2923 ADR: 8,III |
Specification Test Results
Melting point | 64-66° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...