MATR003163

003163-1G
101776-520EA 35.31 USD
101776-520
MATR003163
Purity: 96%
CAS: 585-32-0
MDL Number: MFCD00134680
Molecular Formula: C9H13N
Molecular Weight: 135,21
Boiling Point: 197°C
Hazard: Corrosive
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR