Perfluorocyclobutane ≥99%

Supplier: Matrix Scientific

003121-100G 003121-25G
101776-512EA 846.15 USD
101776-512 101776-510
Perfluorocyclobutane ≥99%
Perfluorocyclobutane

TCA listed: T
Hazard: Nonflammable Gas

Formula: C₄F₈
MW: 200.03 g/mol
Boiling Pt: –6 °C
Melting Pt: –41.4 °C
CAS Number: 115-25-3

Order Now

Specification Test Results

Boiling point -6°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR