1,1,1-Trichlorotrifluoroacetone ≥97%

Supplier: Matrix Scientific

003106-25G
101776-508EA 675.38 USD
101776-508
1,1,1-Trichlorotrifluoroacetone ≥97%
1,1,1-Trichlorotrifluoroacetone

Hazard: Toxic/Irritant

Formula: Cl₃CC(O)CF₃
MW: 215.39 g/mol
Boiling Pt: 86…89 °C
MDL Number: MFCD00042128
CAS Number: 758-42-9

Order Now

Specification Test Results

Boiling point 86-89°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR