Difluoroacetic anhydride ≥95%

Supplier: Matrix Scientific

003068-25G 003068-100G
101776-500EA 264.62 USD
101776-500 101776-498
Difluoroacetic anhydride ≥95%
Difluoroacetic anhydride

Hazard: Corrosive

Formula: C₄H₂F₄O₃
Density: 1.4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Density 1.4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR