Methyl-3-isothiocyanatobenzoate ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(Methoxycarbonyl)phenyl isothiocyanate

003062-5G 003062-1G
101776-492EA 300 USD
101776-492 101776-490
Methyl-3-isothiocyanatobenzoate ≥98%
Methyl 3-isothiocyanatobenzoate

Hazard: Toxic/Refrigerate/Lachrymator

Formula: C₉H₇NO₂S
Boiling Pt: 105 °C (0.45 mmHg)
Density: 1.23
MDL Number: MFCD00041080
CAS Number: 3125-66-4

Order Now

Specification Test Results

Boiling point 105°/0.45m
Density 1.23

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR