2-Chloro-6-fluorotoluene ≥97%

Supplier: Matrix Scientific

002782-100G 002782-500G
101776-376EA 90.77 USD
101776-376 101776-374
2-Chloro-6-fluorotoluene ≥97%
2-Chloro-6-fluorotoluene

TCA listed: T
Hazard: Flammable

Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 154…156 °C
Density: 1.191
Flash Pt: 51 °C
MDL Number: MFCD00000570
CAS Number: 443-83-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 154-156°
Density 1.191
ND20 1.503

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR