2-Chloro-6-fluoro-3-methylbenzylamine

Supplier: Matrix Scientific

002758-1G
101776-342EA 146.15 USD
101776-342
2-Chloro-6-fluoro-3-methylbenzylamine
2-Chloro-6-fluoro-3-methylbenzylamine

Hazard: Corrosive/Stored under Argon/Air Sensitive

Formula: C₈H₉ClFN
MDL Number: MFCD01631395
CAS Number: 261762-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR