2-Chloro-6-fluoro-3-toluoyl chloride
Supplier: Matrix Scientific
002756-1G
002756-5G
101776-338EA
30.77
USD
101776-338
101776-340
2-Chloro-6-fluoro-3-toluoyl chloride
2-Chloro-6-fluoro-3-toluoyl chloride
Hazard: Corrosive
Formula:
C₈H₅Cl₂FO MW: 207.03 g/mol |
MDL Number:
MFCD01631361 CAS Number: 261762-82-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...