1-Phenyl-5-(trifluoromethyl)pyrazole-4-carbonylchloride

Supplier: Matrix Scientific

002669-5G 002669-1G
101776-268EA 110.77 USD
101776-268 101776-270
1-Phenyl-5-(trifluoromethyl)pyrazole-4-carbonylchloride
1-Phenyl-5-(trifluoromethyl)pyrazole-4-carbonylchloride

Hazard: Corrosive

Formula: C₁₁H₆ClF₃N₂O
MW: 274.63 g/mol
Boiling Pt: 110 °C (0.2 mmHg)
Melting Pt: 40…41 °C
MDL Number: MFCD00068139
CAS Number: 175137-14-1

Order Now

Specification Test Results

Boiling point 110°/0.2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR