3-Fluoro-p-toluonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-4-methylbenzonitrile

002569-25G
101776-188EA 50.77 USD
101776-188
3-Fluoro-p-toluonitrile ≥97%
3-Fluoro-p-toluonitrile

Hazard: Toxic

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 204 °C
Melting Pt: 48…50 °C
MDL Number: MFCD00153174
CAS Number: 170572-49-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 204°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR