2-Fluoro-4-(trifluoromethyl)benzonitrile ≥97%
Supplier: Matrix Scientific
002421-1G
002421-5G
101776-060EA
13.85
USD
101776-060
101776-062
2-Fluoro-4-(trifluoromethyl)benzonitrile ≥97%
2-Fluoro-4-(trifluoromethyl)benzonitrile
Hazard: Toxic/Irritant
Formula:
C₈H₃F₄N MW: 189.11 g/mol |
MDL Number:
MFCD00061279 CAS Number: 146070-34-0 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...