2-Methyl-4,4,4-trifluorobutanol ≥95%

Supplier: Matrix Scientific

002410-1G 002410-5G
101776-048EA 121.54 USD
101776-048 101776-050
2-Methyl-4,4,4-trifluorobutanol ≥95%
2-Methyl-4,4,4-trifluorobutanol

Hazard: Flammable

Formula: C₅H₉F₃O
Boiling Pt: 134 °C
MDL Number: MFCD00190639
CAS Number: 107103-95-7

Order Now

Specification Test Results

Boiling point 134°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR