2-Methyl-3-nitrobenzotrifluoride ≥98%
Supplier: Matrix Scientific
002391-5G
002391-25G
102832-746EA
96.92
USD
102832-746
101776-044
2-Methyl-3-nitrobenzotrifluoride ≥98%
2-Methyl-3-nitrobenzotrifluoride
Hazard: Toxic
Formula:
C₈H₆F₃NO₂ MW: 205.14 g/mol Boiling Pt: 86 °C (10 mmHg) |
MDL Number:
MFCD00042322 CAS Number: 6656-49-1 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 86°/10mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...