3-Chloro-3-(4-fluorophenyl)acrylonitrile ≥97%

Supplier: Matrix Scientific

002272-1G
101775-914EA 86.15 USD
101775-914
3-Chloro-3-(4-fluorophenyl)acrylonitrile ≥97%
3-Chloro-3-(4-fluorophenyl)acrylonitrile

Hazard: Toxic

Formula: C₉H₅ClFN
Melting Pt: 90…93 °C
MDL Number: MFCD00052069
CAS Number: 126417-76-3

Order Now

Specification Test Results

Melting point 90-93°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR