2-Chloro-3,3,3-trifluoropropene ≥98%
Supplier: Matrix Scientific
002250-25G
002250-5G
101775-888EA
212.31
USD
101775-888
101775-886
2-Chloro-3,3,3-trifluoropropene ≥98%
2-Chloro-3,3,3-trifluoropropene
Hazard: Nonflammable Gas/Store in Freezer
Formula:
C₃H₂ClF₃ MW: 130.5 g/mol Boiling Pt: 14…15 °C |
MDL Number:
MFCD00082613 CAS Number: 2730-62-3 |
Specification Test Results
Boiling point | 14-15° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...